Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTCACTATTTGGGAGGGAGAGT[A/G]TCTGCTGAAGTAACCTTCTACCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614586 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZDHHC5 - zinc finger DHHC-type containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015457.2 | Intron | NP_056272.2 | ||||
XM_011544899.1 | Intron | XP_011543201.1 | ||||
XM_011544900.1 | Intron | XP_011543202.1 | ||||
XM_011544901.1 | Intron | XP_011543203.1 | ||||
XM_017017498.1 | Intron | XP_016872987.1 |