Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606203 | ||||||||||||||||||||
Literature Links: |
GAB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAB2 - GRB2 associated binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012296.3 | 4826 | UTR 3 | NP_036428.1 | |||
NM_080491.2 | 4826 | UTR 3 | NP_536739.1 | |||
XM_006718753.2 | 4826 | UTR 3 | XP_006718816.1 | |||
XM_011545408.2 | 4826 | UTR 3 | XP_011543710.1 |
USP35 - ubiquitin specific peptidase 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020798.2 | 4826 | Intron | NP_065849.1 | |||
XM_011545187.2 | 4826 | Intron | XP_011543489.1 | |||
XM_011545188.2 | 4826 | Intron | XP_011543490.1 | |||
XM_017018052.1 | 4826 | Intron | XP_016873541.1 | |||
XM_017018053.1 | 4826 | Intron | XP_016873542.1 |