Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606203 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GAB2 - GRB2 associated binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012296.3 | 4415 | UTR 3 | NP_036428.1 | |||
NM_080491.2 | 4415 | UTR 3 | NP_536739.1 | |||
XM_006718753.2 | 4415 | UTR 3 | XP_006718816.1 | |||
XM_011545408.2 | 4415 | UTR 3 | XP_011543710.1 |
USP35 - ubiquitin specific peptidase 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020798.2 | 4415 | Intron | NP_065849.1 | |||
XM_011545187.2 | 4415 | Intron | XP_011543489.1 | |||
XM_011545188.2 | 4415 | Intron | XP_011543490.1 | |||
XM_017018052.1 | 4415 | Intron | XP_016873541.1 | |||
XM_017018053.1 | 4415 | Intron | XP_016873542.1 |
Set Membership: |
HapMap |