Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTCCCCTCCTTTTGCTCAAGAA[C/T]TCGTCCATTCCTTCTCCAACTCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609316 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM31 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRIM31 - tripartite motif containing 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007028.4 | 1607 | UTR 3 | NP_008959.3 | |||
XM_011514264.1 | 1607 | UTR 3 | XP_011512566.1 | |||
XM_011514265.1 | 1607 | Intron | XP_011512567.1 | |||
XM_011514266.1 | 1607 | Intron | XP_011512568.1 |
TRIM31-AS1 - TRIM31 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |