Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGACTCAAGACGCCAACGGCGC[C/G]GTCTTCCTGGGGCCCCAGGGCCTGC
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 601266 | |||||||||||||||||||||||||||||
Literature Links: |
DUT PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
DUT - deoxyuridine triphosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025248.1 | Intron | NP_001020419.1 | ||||
NM_001025249.1 | Intron | NP_001020420.1 | ||||
NM_001948.3 | Intron | NP_001939.1 | ||||
XM_011521315.2 | Intron | XP_011519617.1 | ||||
XM_017021988.1 | Intron | XP_016877477.1 |
Set Membership: |
HapMap |