Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR3G PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POLR3G - RNA polymerase III subunit G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006467.2 | 297 | Intron | NP_006458.2 | |||
XM_005248404.3 | 297 | Intron | XP_005248461.1 | |||
XM_011543100.2 | 297 | Intron | XP_011541402.1 | |||
XM_011543101.2 | 297 | Intron | XP_011541403.1 | |||
XM_011543102.2 | 297 | Intron | XP_011541404.1 | |||
XM_011543103.2 | 297 | Intron | XP_011541405.1 | |||
XM_011543104.2 | 297 | Intron | XP_011541406.1 | |||
XM_011543105.2 | 297 | Intron | XP_011541407.1 | |||
XM_017008957.1 | 297 | UTR 5 | XP_016864446.1 |
Set Membership: |
HapMap |