Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATTGCCTAAATTATTGGCAATT[G/T]GTGAATTGCTGATTGTGTCTGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606830 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGTPBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AGTPBP1 - ATP/GTP binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286715.1 | 4970 | Intron | NP_001273644.1 | |||
NM_001286717.1 | 4970 | Intron | NP_001273646.1 | |||
NM_015239.2 | 4970 | Intron | NP_056054.2 | |||
XM_005251847.3 | 4970 | Intron | XP_005251904.1 | |||
XM_005251848.2 | 4970 | Intron | XP_005251905.1 | |||
XM_005251849.3 | 4970 | Intron | XP_005251906.1 | |||
XM_011518418.2 | 4970 | UTR 3 | XP_011516720.1 | |||
XM_011518420.2 | 4970 | Intron | XP_011516722.1 | |||
XM_011518421.2 | 4970 | Intron | XP_011516723.1 | |||
XM_017014544.1 | 4970 | Intron | XP_016870033.1 | |||
XM_017014545.1 | 4970 | UTR 3 | XP_016870034.1 | |||
XM_017014546.1 | 4970 | Intron | XP_016870035.1 | |||
XM_017014547.1 | 4970 | UTR 3 | XP_016870036.1 | |||
XM_017014548.1 | 4970 | Intron | XP_016870037.1 | |||
XM_017014549.1 | 4970 | UTR 3 | XP_016870038.1 | |||
XM_017014550.1 | 4970 | Intron | XP_016870039.1 |
Set Membership: |
HapMap |