Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAATCTTTTTTTATTATTAATGC[A/G]CACTTTTATATGAATAGTGGTAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616156 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAR2 - fatty acyl-CoA reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271783.1 | Intron | NP_001258712.1 | ||||
NM_001271784.1 | Intron | NP_001258713.1 | ||||
NM_018099.4 | Intron | NP_060569.3 | ||||
XM_011520747.2 | Intron | XP_011519049.1 | ||||
XM_011520748.2 | Intron | XP_011519050.1 | ||||
XM_017019624.1 | Intron | XP_016875113.1 | ||||
XM_017019625.1 | Intron | XP_016875114.1 |