Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603499 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFRSF11A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TNFRSF11A - TNF receptor superfamily member 11a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270949.1 | Intron | NP_001257878.1 | ||||
NM_001270950.1 | Intron | NP_001257879.1 | ||||
NM_001270951.1 | Intron | NP_001257880.1 | ||||
NM_001278268.1 | Intron | NP_001265197.1 | ||||
NM_003839.3 | Intron | NP_003830.1 | ||||
XM_011526244.2 | Intron | XP_011524546.1 | ||||
XM_011526245.2 | Intron | XP_011524547.1 | ||||
XM_017026064.1 | Intron | XP_016881553.1 | ||||
XM_017026065.1 | Intron | XP_016881554.1 | ||||
XM_017026066.1 | Intron | XP_016881555.1 |
Set Membership: |
HapMap |