Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCATAGGAGTTTTTTATTCAGGC[C/T]GCAAAATTCCTGTGTTTTATTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156570 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTR - 5-methyltetrahydrofolate-homocysteine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000254.2 | Intron | NP_000245.2 | ||||
NM_001291939.1 | Intron | NP_001278868.1 | ||||
NM_001291940.1 | Intron | NP_001278869.1 | ||||
XM_005273141.4 | Intron | XP_005273198.1 | ||||
XM_006711770.2 | Intron | XP_006711833.1 | ||||
XM_011544194.2 | Intron | XP_011542496.1 | ||||
XM_017001329.1 | Intron | XP_016856818.1 | ||||
XM_017001330.1 | Intron | XP_016856819.1 |