Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601481 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYO10 - myosin X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012334.2 | 6160 | UTR 3 | NP_036466.2 | |||
XM_005248306.4 | 6160 | UTR 3 | XP_005248363.1 | |||
XM_005248307.2 | 6160 | UTR 3 | XP_005248364.1 | |||
XM_006714475.2 | 6160 | UTR 3 | XP_006714538.1 | |||
XM_011514046.2 | 6160 | UTR 3 | XP_011512348.1 |
Set Membership: |
HapMap |