Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194543 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF69 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF69 - zinc finger protein 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321163.1 | Intron | NP_001308092.1 | ||||
NM_021915.3 | Intron | NP_068734.1 | ||||
XM_006722873.3 | Intron | XP_006722936.1 | ||||
XM_006722874.3 | Intron | XP_006722937.1 | ||||
XM_017027231.1 | Intron | XP_016882720.1 |
ZNF700 - zinc finger protein 700 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271848.1 | Intron | NP_001258777.1 | ||||
NM_144566.2 | Intron | NP_653167.1 |
Set Membership: |
HapMap |