Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL22 - methyltransferase like 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024109.3 | Intron | NP_077014.3 | ||||
XM_005255570.3 | Intron | XP_005255627.1 | ||||
XM_005255571.2 | Intron | XP_005255628.1 | ||||
XM_011522657.2 | Intron | XP_011520959.1 | ||||
XM_017023659.1 | Intron | XP_016879148.1 | ||||
XM_017023660.1 | Intron | XP_016879149.1 | ||||
XM_017023661.1 | Intron | XP_016879150.1 | ||||
XM_017023662.1 | Intron | XP_016879151.1 |