Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTACACATTAAGGCTGGACCCC[A/C]GCGCCCGCACCTATTGGTATGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
70 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609607 MIM: 613466 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHDC9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLHDC9 - kelch domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007255.2 | 656 | Missense Mutation | AGC,CGC | S,R 171 | NP_001007256.1 | |
NM_152366.4 | 656 | Missense Mutation | AGC,CGC | S,R 171 | NP_689579.3 |
NECTIN4 - nectin cell adhesion molecule 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFDN2 - prefoldin subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |