Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604014 MIM: 600923 MIM: 604729 | ||||||||||||||||||||
Literature Links: |
B4GALT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B4GALT3 - beta-1,4-galactosyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199873.1 | 1636 | UTR 3 | NP_001186802.1 | |||
NM_001199874.1 | 1636 | UTR 3 | NP_001186803.1 | |||
NM_003779.3 | 1636 | UTR 3 | NP_003770.1 | |||
XM_005245566.1 | 1636 | UTR 3 | XP_005245623.1 | |||
XM_011510093.1 | 1636 | UTR 3 | XP_011508395.1 | |||
XM_017002714.1 | 1636 | UTR 3 | XP_016858203.1 | |||
XM_017002715.1 | 1636 | Intron | XP_016858204.1 |
USP21 - ubiquitin specific peptidase 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |