Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCCAAACTGAGGCCAAGCCTGC[C/G]AATACTCTCAGCAGCAGTTTTCTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600682 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC16A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC16A1 - solute carrier family 16 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166496.1 | 3742 | UTR 3 | NP_001159968.1 | |||
NM_003051.3 | 3742 | UTR 3 | NP_003042.3 | |||
XM_011542026.2 | 3742 | Intron | XP_011540328.1 | |||
XM_011542027.2 | 3742 | Intron | XP_011540329.1 |
Set Membership: |
HapMap |