Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGGCAAGGGTGGTGGATGTTCA[C/G]AGTGTGACTGCAGAGGGCTTCTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607465 MIM: 614642 | ||||||||||||||||||||
Literature Links: |
CDAN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDAN1 - codanin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138477.2 | Intron | NP_612486.2 | ||||
XM_005254176.4 | Intron | XP_005254233.1 | ||||
XM_011521270.2 | Intron | XP_011519572.1 | ||||
XM_011521271.2 | Intron | XP_011519573.1 | ||||
XM_011521274.2 | Intron | XP_011519576.1 |
STARD9 - StAR related lipid transfer domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |