Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179617 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD51 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAD51 - RAD51 recombinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164269.1 | Intron | NP_001157741.1 | ||||
NM_001164270.1 | Intron | NP_001157742.1 | ||||
NM_002875.4 | Intron | NP_002866.2 | ||||
NM_133487.3 | Intron | NP_597994.3 | ||||
XM_006720626.3 | Intron | XP_006720689.1 | ||||
XM_011521857.2 | Intron | XP_011520159.1 | ||||
XM_011521858.2 | Intron | XP_011520160.1 | ||||
XM_011521859.2 | Intron | XP_011520161.1 | ||||
XM_011521860.2 | Intron | XP_011520162.1 | ||||
XM_011521861.2 | Intron | XP_011520163.1 | ||||
XM_011521862.2 | Intron | XP_011520164.1 |
RAD51-AS1 - RAD51 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |