Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCCTGTTTCTTTGCTTTTACAT[C/G]TGAAAATATTAACATTTGTTAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611246 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAL - adenosine deaminase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012969.3 | Intron | NP_001012987.1 | ||||
NM_001159280.2 | Intron | NP_001152752.1 | ||||
NM_001324364.1 | Intron | NP_001311293.1 | ||||
NM_001324365.1 | Intron | NP_001311294.1 | ||||
NM_001324366.1 | Intron | NP_001311295.1 | ||||
NM_001324367.1 | Intron | NP_001311296.1 | ||||
NM_001324368.1 | Intron | NP_001311297.1 | ||||
XM_011521291.2 | Intron | XP_011519593.1 | ||||
XM_017021964.1 | Intron | XP_016877453.1 | ||||
XM_017021965.1 | Intron | XP_016877454.1 | ||||
XM_017021966.1 | Intron | XP_016877455.1 | ||||
XM_017021967.1 | Intron | XP_016877456.1 | ||||
XM_017021968.1 | Intron | XP_016877457.1 | ||||
XM_017021969.1 | Intron | XP_016877458.1 |
LCMT2 - leucine carboxyl methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |