Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGTTGATGCAGTGTTGGTATGT[A/G]CGGCCATGTTAGATGTTAGAACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601203 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1RL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL1RL1 - interleukin 1 receptor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282408.1 | Intron | NP_001269337.1 | ||||
NM_003856.3 | Intron | NP_003847.2 | ||||
NM_016232.4 | Intron | NP_057316.3 | ||||
XM_006712839.3 | Intron | XP_006712902.1 | ||||
XM_011512151.1 | Intron | XP_011510453.1 |
Set Membership: |
HapMap |