Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTCAGAGGTGGTTTGATGTCAG[C/T]AGTGAGACTGTTTGCACCCTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604410 MIM: 605640 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928517 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928517 - uncharacterized LOC101928517 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGLEC7 - sialic acid binding Ig like lectin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277201.1 | Intron | NP_001264130.1 | ||||
NM_014385.3 | Intron | NP_055200.1 | ||||
NM_016543.3 | Intron | NP_057627.2 | ||||
XM_011526721.2 | Intron | XP_011525023.1 |
SIGLEC9 - sialic acid binding Ig like lectin 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |