Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTATTTTTTCTTTTGTTACTTA[G/T]GCTTTTGGTGTCATAGCTAAGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6806 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6806 - microRNA 6806 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN22 - zinc finger and SCAN domain containing 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321116.1 | Intron | NP_001308045.1 | ||||
NM_001321117.1 | Intron | NP_001308046.1 | ||||
NM_181846.2 | Intron | NP_862829.1 | ||||
XM_006723192.3 | Intron | XP_006723255.1 | ||||
XM_011526917.2 | Intron | XP_011525219.1 | ||||
XM_011526918.2 | Intron | XP_011525220.1 | ||||
XM_017026746.1 | Intron | XP_016882235.1 | ||||
XM_017026747.1 | Intron | XP_016882236.1 |