Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610730 MIM: 600940 | ||||||||||||||||||||
Literature Links: |
CCT6B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCT6B - chaperonin containing TCP1 subunit 6B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIG3 - DNA ligase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002311.4 | 217 | Missense Mutation | CGT,TGT | R,C 30 | NP_002302.2 | |
NM_013975.3 | 217 | Missense Mutation | CGT,TGT | R,C 30 | NP_039269.2 | |
XM_005257970.3 | 217 | Missense Mutation | CGT,TGT | R,C 39 | XP_005258027.1 | |
XM_006721896.3 | 217 | Missense Mutation | CGT,TGT | R,C 39 | XP_006721959.1 | |
XM_017024624.1 | 217 | Missense Mutation | CGT,TGT | R,C 30 | XP_016880113.1 |