Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGTCCTGCCCAGCGTGGCTCTT[C/G]TGATGGCTTCCGAGCCCCTGGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605855 | ||||||||||||||||||||
Literature Links: |
ATP10A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP10A - ATPase phospholipid transporting 10A (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024490.3 | Intron | NP_077816.1 | ||||
XM_005268261.4 | Intron | XP_005268318.1 | ||||
XM_011521826.2 | Intron | XP_011520128.1 | ||||
XM_011521828.2 | Intron | XP_011520130.1 | ||||
XM_011521829.2 | Intron | XP_011520131.1 | ||||
XM_011521830.2 | Intron | XP_011520132.2 | ||||
XM_017022436.1 | Intron | XP_016877925.1 | ||||
XM_017022437.1 | Intron | XP_016877926.1 |