Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTAAGTAGGGGAGGAAAGAGTT[A/G]AGGAGGGAACATTGATTGGGTTATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180240 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371934 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105371934 - uncharacterized LOC105371934 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RARA - retinoic acid receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000964.3 | Intron | NP_000955.1 | ||||
NM_001024809.3 | Intron | NP_001019980.1 | ||||
NM_001145301.2 | Intron | NP_001138773.1 | ||||
NM_001145302.2 | Intron | NP_001138774.1 | ||||
XM_005257552.4 | Intron | XP_005257609.1 | ||||
XM_005257553.1 | Intron | XP_005257610.1 | ||||
XM_005257554.1 | Intron | XP_005257611.1 | ||||
XM_011525095.1 | Intron | XP_011523397.1 | ||||
XM_011525096.1 | Intron | XP_011523398.1 | ||||
XM_017024920.1 | Intron | XP_016880409.1 |
Set Membership: |
HapMap |