Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604839 MIM: 612548 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FKBP6 - FK506 binding protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135211.2 | Intron | NP_001128683.1 | ||||
NM_001281304.1 | Intron | NP_001268233.1 | ||||
NM_003602.4 | Intron | NP_003593.3 | ||||
XM_006716153.2 | Intron | XP_006716216.1 | ||||
XM_017012741.1 | Intron | XP_016868230.1 | ||||
XM_017012742.1 | Intron | XP_016868231.1 |
TRIM50 - tripartite motif containing 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |