Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAHD2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAHD2B - fumarylacetoacetate hydrolase domain containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320848.1 | Intron | NP_001307777.1 | ||||
NM_001320849.1 | Intron | NP_001307778.1 | ||||
NM_199336.2 | Intron | NP_955368.1 | ||||
XM_011510745.2 | Intron | XP_011509047.1 | ||||
XM_011510746.2 | Intron | XP_011509048.1 | ||||
XM_011510747.2 | Intron | XP_011509049.1 | ||||
XM_011510748.2 | Intron | XP_011509050.1 | ||||
XM_011510750.2 | Intron | XP_011509052.1 | ||||
XM_017003470.1 | Intron | XP_016858959.1 | ||||
XM_017003471.1 | Intron | XP_016858960.1 | ||||
XM_017003472.1 | Intron | XP_016858961.1 | ||||
XM_017003473.1 | Intron | XP_016858962.1 |
LOC653924 - glycerol-3-phosphate acyltransferase 2, mitochondrial pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |