Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACGGTCAAAACAATGCGTCTGGA[T/C]GGGCAGCATTGTTTCTTATTGTAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604336 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105378260 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105378260 - histone-lysine N-methyltransferase 2C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPTE - transmembrane phosphatase with tensin homology | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290224.1 | Intron | NP_001277153.1 | ||||
NM_199259.3 | Intron | NP_954868.1 | ||||
NM_199260.3 | Intron | NP_954869.1 | ||||
NM_199261.3 | Intron | NP_954870.2 |
Set Membership: |
HapMap |