Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614695 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPRD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPRD2 - regulation of nuclear pre-mRNA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297673.1 | Intron | NP_001284602.1 | ||||
NM_001297674.1 | Intron | NP_001284603.1 | ||||
NM_015203.4 | Intron | NP_056018.2 | ||||
XM_005245033.2 | Intron | XP_005245090.1 | ||||
XM_006711240.2 | Intron | XP_006711303.1 | ||||
XM_011509344.2 | Intron | XP_011507646.1 | ||||
XM_017000763.1 | Intron | XP_016856252.1 | ||||
XM_017000764.1 | Intron | XP_016856253.1 | ||||
XM_017000765.1 | Intron | XP_016856254.1 | ||||
XM_017000766.1 | Intron | XP_016856255.1 | ||||
XM_017000767.1 | Intron | XP_016856256.1 |