Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACACGAAAAATCAGAGGTAAAC[A/G]CAGACCTTTTTCTGATTTTGCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604445 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINB13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SERPINB13 - serpin family B member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307923.1 | Intron | NP_001294852.1 | ||||
NM_012397.3 | Intron | NP_036529.1 | ||||
XM_005266707.3 | Intron | XP_005266764.1 | ||||
XM_011526029.2 | Intron | XP_011524331.1 |
Set Membership: |
HapMap |