Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613749 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF260 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF260 - zinc finger protein 260 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012756.2 | 2119 | Silent Mutation | TCA,TCG | S,S 401 | NP_001012774.1 | |
NM_001166036.1 | 2119 | Silent Mutation | TCA,TCG | S,S 401 | NP_001159508.1 | |
NM_001166037.1 | 2119 | Silent Mutation | TCA,TCG | S,S 401 | NP_001159509.1 | |
NM_001166038.1 | 2119 | Silent Mutation | TCA,TCG | S,S 401 | NP_001159510.1 | |
XM_011526897.2 | 2119 | Silent Mutation | TCA,TCG | S,S 401 | XP_011525199.1 | |
XM_017026741.1 | 2119 | Silent Mutation | TCA,TCG | S,S 401 | XP_016882230.1 |
Set Membership: |
HapMap |