Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604144 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC7A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC7A9 - solute carrier family 7 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126335.1 | Intron | NP_001119807.1 | ||||
NM_001243036.1 | Intron | NP_001229965.1 | ||||
NM_014270.4 | Intron | NP_055085.1 | ||||
XM_006722992.1 | Intron | XP_006723055.1 | ||||
XM_011526402.2 | Intron | XP_011524704.1 | ||||
XM_017026230.1 | Intron | XP_016881719.1 |
TDRD12 - tudor domain containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |