Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602783 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPG7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPG7 - SPG7, paraplegin matrix AAA peptidase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003119.3 | Intron | NP_003110.1 | ||||
NM_199367.2 | Intron | NP_955399.1 | ||||
XM_005256321.4 | Intron | XP_005256378.1 | ||||
XM_006721264.3 | Intron | XP_006721327.1 | ||||
XM_017023597.1 | Intron | XP_016879086.1 | ||||
XM_017023598.1 | Intron | XP_016879087.1 | ||||
XM_017023599.1 | Intron | XP_016879088.1 |