Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGGTGCAGGATTTTTCAGGATT[C/G]TGAAATGAGTAGCCTACTGGCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605689 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPNE7 - copine 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014427.4 | Intron | NP_055242.1 | ||||
NM_153636.2 | Intron | NP_705900.1 | ||||
XM_011523000.1 | Intron | XP_011521302.1 | ||||
XM_011523001.2 | Intron | XP_011521303.1 | ||||
XM_017023138.1 | Intron | XP_016878627.1 | ||||
XM_017023139.1 | Intron | XP_016878628.1 | ||||
XM_017023140.1 | Intron | XP_016878629.1 | ||||
XM_017023141.1 | Intron | XP_016878630.1 |