Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTATACTGACCTGGGCCCCAAGT[A/G]AGTAGAATAAAAAGGAGATGTTTTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
PLPPR5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
PLPPR5 - phospholipid phosphatase related 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010861.2 | 4161 | UTR 3 | NP_001010861.1 | |||
NM_001037317.1 | 4161 | UTR 3 | NP_001032394.1 | |||
XM_011540836.2 | 4161 | UTR 3 | XP_011539138.1 | |||
XM_011540837.1 | 4161 | Intron | XP_011539139.1 | |||
XM_011540838.2 | 4161 | UTR 3 | XP_011539140.1 |
Set Membership: |
HapMap |