Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGACCTCGGTGCCCAAGTGCT[C/G]CCTCAGTGGCCTCCTGAGTCTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611877 MIM: 615628 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAIAP2L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAIAP2L1 - BAI1 associated protein 2 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BRI3 - brain protein I3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159491.1 | Intron | NP_001152963.1 | ||||
NM_015379.4 | Intron | NP_056194.1 | ||||
XM_017011931.1 | Intron | XP_016867420.1 | ||||
XM_017011932.1 | Intron | XP_016867421.1 | ||||
XM_017011933.1 | Intron | XP_016867422.1 | ||||
XM_017011934.1 | Intron | XP_016867423.1 | ||||
XM_017011935.1 | Intron | XP_016867424.1 | ||||
XM_017011936.1 | Intron | XP_016867425.1 |