Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600079 | ||||||||||||||||||||
Literature Links: |
PTPN12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPN12 - protein tyrosine phosphatase, non-receptor type 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131008.1 | 131 | Intron | NP_001124480.1 | |||
NM_001131009.1 | 131 | Intron | NP_001124481.1 | |||
NM_002835.3 | 131 | Missense Mutation | ATC,GTC | I,V 14 | NP_002826.3 | |
XM_005250518.2 | 131 | UTR 5 | XP_005250575.1 | |||
XM_006716073.3 | 131 | Missense Mutation | ATC,GTC | I,V 14 | XP_006716136.1 | |
XM_011516444.1 | 131 | UTR 5 | XP_011514746.1 | |||
XM_017012474.1 | 131 | Intron | XP_016867963.1 |