Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTAAAAAAAACCATCATTTCCT[A/G]TAAGATAAACAACACAATTTCCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300562 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLITRK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLITRK4 - SLIT and NTRK like family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184749.2 | 7856 | UTR 3 | NP_001171678.1 | |||
NM_001184750.2 | 7856 | UTR 3 | NP_001171679.1 | |||
NM_173078.4 | 7856 | UTR 3 | NP_775101.1 | |||
XM_005262365.4 | 7856 | Intron | XP_005262422.1 | |||
XM_011531265.2 | 7856 | Intron | XP_011529567.1 |