Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATATATATATATATATAATGCGG[G/T]TAAGCTTACTGCTCGTGACATTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610747 | ||||||||||||||||||||
Literature Links: |
SAMD4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SAMD4A - sterile alpha motif domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161576.2 | Intron | NP_001155048.2 | ||||
NM_001161577.1 | Intron | NP_001155049.1 | ||||
NM_015589.5 | Intron | NP_056404.4 |