Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGTTAAACAGAGGACAGTATTG[G/T]TATGAGTCCTGATAGATAGGACTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616761 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUSD6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SUSD6 - sushi domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014734.3 | Intron | NP_055549.1 | ||||
XM_011537412.1 | Intron | XP_011535714.1 | ||||
XM_011537413.1 | Intron | XP_011535715.1 | ||||
XM_017021838.1 | Intron | XP_016877327.1 | ||||
XM_017021839.1 | Intron | XP_016877328.1 |