Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603210 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JRK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
JRK - Jrk helix-turn-helix protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077527.2 | 11798 | Intron | NP_001070995.2 | |||
NM_001279352.1 | 11798 | Intron | NP_001266281.1 | |||
NM_003724.3 | 11798 | Intron | NP_003715.3 | |||
XM_006716677.3 | 11798 | UTR 3 | XP_006716740.1 | |||
XM_006716678.3 | 11798 | UTR 3 | XP_006716741.1 | |||
XM_011517354.1 | 11798 | Intron | XP_011515656.1 | |||
XM_011517355.1 | 11798 | Intron | XP_011515657.1 |
Set Membership: |
HapMap |