Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCTGTACTTGCCAGTTTTGGGG[C/T]CTCAACTAATACACCCACAAACTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATAD2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GATAD2A - GATA zinc finger domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300946.1 | Intron | NP_001287875.1 | ||||
NM_017660.3 | Intron | NP_060130.3 | ||||
XM_011528105.1 | Intron | XP_011526407.1 | ||||
XM_017026901.1 | Intron | XP_016882390.1 | ||||
XM_017026902.1 | Intron | XP_016882391.1 | ||||
XM_017026903.1 | Intron | XP_016882392.1 | ||||
XM_017026904.1 | Intron | XP_016882393.1 | ||||
XM_017026905.1 | Intron | XP_016882394.1 | ||||
XM_017026906.1 | Intron | XP_016882395.1 | ||||
XM_017026907.1 | Intron | XP_016882396.1 | ||||
XM_017026908.1 | Intron | XP_016882397.1 | ||||
XM_017026909.1 | Intron | XP_016882398.1 | ||||
XM_017026910.1 | Intron | XP_016882399.1 | ||||
XM_017026911.1 | Intron | XP_016882400.1 |