Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300359 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CT45A10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CT45A10 - cancer/testis antigen family 45, member A10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAGE1 - sarcoma antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018666.2 | Intron | NP_061136.2 | ||||
XM_017029621.1 | Intron | XP_016885110.1 | ||||
XM_017029622.1 | Intron | XP_016885111.1 | ||||
XM_017029623.1 | Intron | XP_016885112.1 | ||||
XM_017029624.1 | Intron | XP_016885113.1 | ||||
XM_017029625.1 | Intron | XP_016885114.1 | ||||
XM_017029626.1 | Intron | XP_016885115.1 | ||||
XM_017029627.1 | Intron | XP_016885116.1 |
Set Membership: |
HapMap |