Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610555 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1GALT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1GALT1 - core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020156.4 | Intron | NP_064541.1 | ||||
XM_005249812.1 | Intron | XP_005249869.1 | ||||
XM_011515453.2 | Intron | XP_011513755.1 | ||||
XM_011515455.2 | Intron | XP_011513757.1 | ||||
XM_011515456.2 | Intron | XP_011513758.1 | ||||
XM_017012442.1 | Intron | XP_016867931.1 | ||||
XM_017012443.1 | Intron | XP_016867932.1 | ||||
XM_017012444.1 | Intron | XP_016867933.1 | ||||
XM_017012445.1 | Intron | XP_016867934.1 | ||||
XM_017012446.1 | Intron | XP_016867935.1 | ||||
XM_017012447.1 | Intron | XP_016867936.1 | ||||
XM_017012448.1 | Intron | XP_016867937.1 | ||||
XM_017012449.1 | Intron | XP_016867938.1 |
Set Membership: |
HapMap |