Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601423 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf73 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C12orf73 - chromosome 12 open reading frame 73 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TDG - thymine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003211.4 | Intron | NP_003202.3 | ||||
XM_005269125.1 | Intron | XP_005269182.1 | ||||
XM_011538714.1 | Intron | XP_011537016.1 | ||||
XM_011538715.1 | Intron | XP_011537017.1 |
Set Membership: |
HapMap |