Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATTCATGTAATGCAGGATGTAT[A/G]TTGGAGAAGGTTAAGTACAGCCACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142620 MIM: 610434 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD16A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABHD16A - abhydrolase domain containing 16A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177515.1 | 2073 | UTR 3 | NP_001170986.1 | |||
NM_021160.2 | 2073 | UTR 3 | NP_066983.1 |
LY6G5C - lymphocyte antigen 6 complex, locus G5C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |