Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCCGCCTGCTGCAGTGGGAGG[C/T]TGCTCCGAGAAGGCAAAGGGCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615137 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WBSCR17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WBSCR17 - Williams-Beuren syndrome chromosome region 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022479.2 | Intron | NP_071924.1 | ||||
XM_011516467.2 | Intron | XP_011514769.1 | ||||
XM_011516469.2 | Intron | XP_011514771.1 | ||||
XM_017012521.1 | Intron | XP_016868010.1 |
Set Membership: |
HapMap |