Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATTTGTCATGTTGATCATTTT[C/T]TCCTTCACTTTCCTTGGCTTAGTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605671 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMXL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DMXL1 - Dmx like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290321.2 | Intron | NP_001277250.1 | ||||
NM_001290322.2 | Intron | NP_001277251.1 | ||||
NM_005509.5 | Intron | NP_005500.4 | ||||
XM_005271909.4 | Intron | XP_005271966.1 | ||||
XM_005271910.4 | Intron | XP_005271967.1 | ||||
XM_005271912.2 | Intron | XP_005271969.1 | ||||
XM_011543212.2 | Intron | XP_011541514.1 | ||||
XM_011543213.2 | Intron | XP_011541515.1 | ||||
XM_011543214.2 | Intron | XP_011541516.1 | ||||
XM_011543215.2 | Intron | XP_011541517.1 | ||||
XM_017009141.1 | Intron | XP_016864630.1 | ||||
XM_017009142.1 | Intron | XP_016864631.1 | ||||
XM_017009143.1 | Intron | XP_016864632.1 | ||||
XM_017009144.1 | Intron | XP_016864633.1 | ||||
XM_017009145.1 | Intron | XP_016864634.1 | ||||
XM_017009146.1 | Intron | XP_016864635.1 | ||||
XM_017009147.1 | Intron | XP_016864636.1 | ||||
XM_017009148.1 | Intron | XP_016864637.1 |
LOC105379143 - uncharacterized LOC105379143 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |