Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613859 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UMODL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UMODL1 - uromodulin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004416.2 | Intron | NP_001004416.2 | ||||
NM_001199527.1 | Intron | NP_001186456.1 | ||||
NM_001199528.2 | Intron | NP_001186457.2 | ||||
NM_173568.3 | Intron | NP_775839.3 | ||||
XM_011529797.1 | Intron | XP_011528099.1 | ||||
XM_017028506.1 | Intron | XP_016883995.1 | ||||
XM_017028507.1 | Intron | XP_016883996.1 | ||||
XM_017028508.1 | Intron | XP_016883997.1 |
Set Membership: |
HapMap |