Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCTGCAGCATGTGCATTTTTG[G/T]TTTTTTCTGTAAGCTGCATTCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602230 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCA5L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LCA5L - LCA5L, lebercilin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6508 - microRNA 6508 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3BGR - SH3 domain binding glutamate rich protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001713.1 | Intron | NP_001001713.1 | ||||
NM_001317740.1 | Intron | NP_001304669.1 | ||||
NM_001317741.1 | Intron | NP_001304670.1 | ||||
NM_001317742.1 | Intron | NP_001304671.1 | ||||
NM_007341.2 | Intron | NP_031367.1 |
WRB-SH3BGR - WRB-SH3BGR readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317744.1 | Intron | NP_001304673.1 |